Assay Details
| Gene Symbol: Mon2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163024.1 | NP_001156496.1 | 3 - 4 | 851 | 83 | |
| NM_001163025.1 | NP_001156497.1 | 3 - 4 | 851 | 83 | ||
| NM_153395.2 | NP_700444.2 | 3 - 4 | 851 | 83 | ||
| GenBank mRNA | AA542162.1 | - | 3 - 4 | 339 | 83 | |
| AB014059.1 | - | 3 - 4 | 373 | 83 | ||
| AK122425.1 | - | 3 - 4 | 851 | 83 | ||
| AK161217.1 | - | 3 - 4 | 730 | 83 | ||
| BC064131.1 | - | 3 - 4 | 341 | 83 | ||
| BC150943.1 | - | 3 - 4 | 372 | 9087854 | 83 | |
| BE955337.1 | - | 1 - 2 | 49 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mon2
Genomic Map