Assay Details
| Gene Symbol: Mon2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163024.1 | NP_001156496.1 | 22 - 23 | 3114 | 103 | |
| NM_001163025.1 | NP_001156497.1 | 21 - 22 | 3114 | 103 | ||
| NM_153395.2 | NP_700444.2 | 21 - 22 | 3111 | 103 | ||
| GenBank mRNA | AB014059.1 | - | 21 - 22 | 2633 | 103 | |
| AK012182.2 | - | 1 - 2 | 51 | 103 | ||
| AK122425.1 | - | 22 - 23 | 3114 | 103 | ||
| AK191210.1 | - | 1 - 2 | 75 | 103 | ||
| AK194501.1 | - | 1 - 2 | 35 | 103 | ||
| BC048406.1 | - | 9 - 10 | 890 | 103 | ||
| BC055354.1 | - | 4 - 5 | 357 | 103 | ||
| BC150943.1 | - | 21 - 22 | 2635 | 9087854 | 103 | |
| CX224591.1 | - | 6 - 7 | 505 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mon2
Genomic Map