Assay Details
| Gene Symbol: Rbm45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153405.2 | NP_700454.1 | 5 - 6 | 1002 | 100 | |
| GenBank mRNA | AB036992.1 | - | 5 - 6 | 918 | 100 | |
| AK080420.1 | - | 5 - 6 | 976 | 100 | ||
| AY152391.1 | - | 5 - 6 | 1002 | 100 | ||
| BC057890.1 | - | 5 - 6 | 913 | 4979982 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbm45
Genomic Map