Assay Details
| Gene Symbol: Ints9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253731.1 | NP_001240660.1 | 5 - 6 | 752 | 60 | |
| NM_153414.4 | NP_700463.2 | 5 - 6 | 720 | 60 | ||
| GenBank mRNA | AK035769.1 | - | 5 - 6 | 464 | 60 | |
| AK038979.1 | - | 5 - 6 | 453 | 60 | ||
| AK077634.1 | - | 5 - 6 | 721 | 60 | ||
| AK136992.1 | - | 5 - 6 | 447 | 60 | ||
| AK150195.1 | - | 5 - 6 | 447 | 60 | ||
| BC028953.1 | - | 4 - 5 | 410 | 5362016 | 60 | |
| BC055700.1 | - | 5 - 6 | 419 | 6808816 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ints9
Genomic Map