Assay Details
| Gene Symbol: Aaas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153416.2 | NP_700465.2 | 11 - 12 | 1227 | 93 | |
| GenBank mRNA | AK034591.1 | - | 11 - 12 | 1122 | 93 | |
| AK079849.1 | - | 11 - 12 | 1164 | 93 | ||
| AK083401.1 | - | 11 - 12 | 1141 | 93 | ||
| AK083537.1 | - | 11 - 12 | 1148 | 93 | ||
| AK087134.1 | - | 11 - 12 | 1203 | 93 | ||
| AK088247.1 | - | 11 - 12 | 1144 | 93 | ||
| AK134406.1 | - | 11 - 12 | 1118 | 93 | ||
| AK165243.1 | - | 11 - 12 | 1142 | 93 | ||
| AK166118.1 | - | 11 - 12 | 1142 | 93 | ||
| AK169523.1 | - | 11 - 12 | 1179 | 93 | ||
| BC018191.1 | - | 11 - 12 | 1128 | 3994248 | 93 | |
| BC025501.1 | - | 11 - 12 | 1174 | 5321013 | 93 | |
| BC026604.1 | - | 10 - 11 | 1681 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aaas
Genomic Map