Assay Details
| Gene Symbol: Aaas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153416.2 | NP_700465.2 | 13 - 14 | 1387 | 122 | |
| GenBank mRNA | AK034591.1 | - | 13 - 14 | 1282 | 122 | |
| AK079849.1 | - | 12 - 13 | 1395 | 122 | ||
| AK083401.1 | - | 13 - 14 | 1301 | 122 | ||
| AK083537.1 | - | 13 - 14 | 1308 | 122 | ||
| AK087134.1 | - | 13 - 14 | 1363 | 122 | ||
| AK088247.1 | - | 13 - 14 | 1304 | 122 | ||
| AK134406.1 | - | 13 - 14 | 1278 | 122 | ||
| AK165243.1 | - | 13 - 14 | 1302 | 122 | ||
| AK166118.1 | - | 13 - 14 | 1302 | 122 | ||
| AK169523.1 | - | 13 - 14 | 1339 | 122 | ||
| BC018191.1 | - | 13 - 14 | 1288 | 3994248 | 122 | |
| BC025501.1 | - | 13 - 14 | 1334 | 5321013 | 122 | |
| BC026604.1 | - | 12 - 13 | 1841 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aaas
Genomic Map