Assay Details
| Gene Symbol: Aaas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153416.2 | NP_700465.2 | 14 - 15 | 1473 | 79 | |
| GenBank mRNA | AK034591.1 | - | 14 - 15 | 1368 | 79 | |
| AK079849.1 | - | 13 - 14 | 1481 | 79 | ||
| AK083401.1 | - | 14 - 15 | 1387 | 79 | ||
| AK083537.1 | - | 14 - 15 | 1394 | 79 | ||
| AK087134.1 | - | 14 - 15 | 1449 | 79 | ||
| AK088247.1 | - | 14 - 15 | 1390 | 79 | ||
| AK134406.1 | - | 14 - 15 | 1364 | 79 | ||
| AK165243.1 | - | 14 - 15 | 1388 | 79 | ||
| AK166118.1 | - | 14 - 15 | 1388 | 79 | ||
| AK169523.1 | - | 14 - 15 | 1425 | 79 | ||
| BC018191.1 | - | 14 - 15 | 1374 | 3994248 | 79 | |
| BC025501.1 | - | 14 - 15 | 1420 | 5321013 | 79 | |
| BC026604.1 | - | 13 - 14 | 1927 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aaas
Genomic Map