Assay Details
| Gene Symbol: Clstn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153508.4 | NP_705728.1 | 9 - 10 | 1787 | 86 | |
| GenBank mRNA | AJ278486.1 | - | 9 - 10 | 1654 | 86 | |
| AK032336.1 | - | 9 - 10 | 1729 | 86 | ||
| BC041781.1 | - | 9 - 10 | 1486 | 86 | ||
| BC055054.1 | - | 9 - 10 | 1719 | 6493096 | 86 | |
More Information
Set Membership: |
|

Clstn3
Genomic Map