Assay Details
| Gene Symbol: Lrrc45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153545.2 | NP_705773.2 | 5 - 6 | 664 | 71 | |
| GenBank mRNA | AK081566.1 | - | 5 - 6 | 667 | 71 | |
| AK153361.1 | - | 5 - 6 | 665 | 71 | ||
| BC023296.1 | - | 5 - 6 | 644 | 3662961 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc45
Genomic Map