Assay Details
| Gene Symbol: Marf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081154.2 | NP_001074623.1 | 10 - 11 | 2454 | 70 | |
| GenBank mRNA | BC031575.1 | - | 3 - 4 | 626 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Marf1
Genomic Map