Assay Details
| Gene Symbol: Dennd1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153551.1 | NP_705779.1 | 10 - 11 | 788 | 68 | |
| GenBank mRNA | AK040537.1 | - | 10 - 11 | 806 | 68 | |
| BC035208.1 | - | 10 - 11 | 788 | 3671384 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dennd1c
Genomic Map