Assay Details
| Gene Symbol: Thoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153552.3 | NP_705780.1 | 7 - 8 | 530 | 100 | |
| GenBank mRNA | AK031785.1 | - | 7 - 8 | 531 | 100 | |
| AK032200.1 | - | 7 - 8 | 531 | 100 | ||
| AK042867.1 | - | 7 - 8 | 536 | 100 | ||
| AK047991.1 | - | 3 - 4 | 1793 | 100 | ||
| AK052835.1 | - | 7 - 8 | 526 | 100 | ||
| AK139307.1 | - | 7 - 8 | 536 | 100 | ||
| AK167374.1 | - | 7 - 8 | 544 | 100 | ||
| AK168619.1 | - | 7 - 8 | 795 | 100 | ||
| BC024951.1 | - | 7 - 8 | 702 | 4010605 | 100 | |
| BC027149.1 | - | 1 - 2 | 112 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Thoc1
Genomic Map