Assay Details
| Gene Symbol: Dcaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098114.1 | - | 1 - 2 | 173 | 86 | |
| BC023804.1 | - | 1 - 2 | 208 | 5352941 | 86 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK098114.1,BC023804.1 transcripts.
Set Membership: |
|

Dcaf8
Genomic Map