Assay Details
| Gene Symbol: Dcaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153555.2 | NP_705783.1 | 6 - 7 | 1186 | 84 | |
| GenBank mRNA | AK035701.1 | - | 6 - 7 | 1085 | 84 | |
| AK098114.1 | - | 6 - 7 | 1205 | 84 | ||
| AK146825.1 | - | 6 - 7 | 1113 | 84 | ||
| AK162622.1 | - | 6 - 7 | 1208 | 84 | ||
| AK164799.1 | - | 6 - 7 | 1113 | 84 | ||
| AK167059.1 | - | 6 - 7 | 1062 | 84 | ||
| BC023704.1 | - | 7 - 8 | 1127 | 5345701 | 84 | |
| BC023804.1 | - | 7 - 8 | 1240 | 5352941 | 84 | |
| BC078641.1 | - | 6 - 7 | 2098 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf8
Genomic Map