Assay Details
| Gene Symbol: Obp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153558.1 | NP_705786.1 | 2 - 3 | 240 | 83 | |
| GenBank mRNA | AY360148.1 | - | 2 - 3 | 276 | 83 | |
| BC027556.1 | - | 2 - 3 | 240 | 1434737 | 83 | |
More Information
Set Membership: |
|

Obp2a
Genomic Map