Assay Details
| Gene Symbol: Strip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153563.2 | NP_705791.2 | 16 - 17 | 1818 | 89 | |
| GenBank mRNA | AK032065.1 | - | 16 - 17 | 1800 | 89 | |
| AK122553.1 | - | 15 - 16 | 2901 | 89 | ||
| AK148011.1 | - | 16 - 17 | 1803 | 89 | ||
| AK169644.1 | - | 15 - 16 | 1686 | 89 | ||
| BC023952.1 | - | 16 - 17 | 1794 | 5320305 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Strip1
Genomic Map