Assay Details
| Gene Symbol: Gbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153564.2 | NP_705792.2 | 1 - 2 | 58 | 63 | |
| GenBank mRNA | AF422243.1 | - | 1 - 2 | 40 | 63 | |
| AF487898.1 | - | 1 - 2 | 50 | 63 | ||
| AK030414.1 | - | 1 - 2 | 58 | 63 | ||
| BC058555.1 | - | 1 - 2 | 92 | 5293575 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF422243.1, AF487898.1, AK030414.1, BC058555.1, NM_153564.2 transcripts.
Set Membership: |
|

Gbp5
Genomic Map