Assay Details
| Gene Symbol: Yrdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153566.2 | NP_705794.2 | 2 - 3 | 528 | 70 | |
| GenBank mRNA | AK153627.1 | - | 2 - 3 | 528 | 70 | |
| AY283537.1 | - | 2 - 3 | 504 | 70 | ||
| BC023823.1 | - | 2 - 3 | 514 | 5343817 | 70 | |
More Information
Set Membership: |
|

Yrdc
Genomic Map