Assay Details
| Gene Symbol: Slain2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153567.3 | NP_705795.2 | 7 - 8 | 1708 | 103 | |
| GenBank mRNA | BC019454.1 | - | 1 - 2 | 103 | ||
| BC038019.1 | - | 5 - 6 | 913 | 3597423 | 103 | |
| CK619668.1 | - | 3 - 4 | 415 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slain2
Genomic Map