Assay Details
| Gene Symbol: Hscb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153571.2 | NP_705799.2 | 4 - 5 | 611 | 85 | |
| GenBank mRNA | AK131961.1 | - | 4 - 5 | 613 | 85 | |
| BC027641.1 | - | 4 - 5 | 575 | 1244839 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hscb
Genomic Map