Assay Details
| Gene Symbol: Cxcl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153576.2 | NP_705804.2 | 1 - 2 | 188 | 94 | |
| GenBank mRNA | AK143005.1 | - | 1 - 2 | 104 | 94 | |
| AY374329.1 | - | 1 - 2 | 91 | 94 | ||
| AY598463.1 | - | 1 - 2 | 189 | 94 | ||
| BC024561.1 | - | 1 - 2 | 188 | 5004999 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25010336 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25010335 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2006722 | NM_153576.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2006722 | AK143005.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2006722 | BC024561.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843057 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523342 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 968739 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 250629 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 232711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572587 | BC038648.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572587 | XM_001475348.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572587 | XM_901330.3 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 826977 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 140523 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544927 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132044 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138043 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2006722 | NM_153576.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2006722 | AK143005.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2006722 | BC024561.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010282 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06143574 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138043 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102601 | - | - |
| Whole Mouse Genome G4122A | A_51_P294235 | NM_153576.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294235 | BC024561.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138191 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138043 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102601 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06307098 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06442268 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06307097 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06307095 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06084733 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25010335 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25010336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06084735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06354981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06084733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05970787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06307098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05960133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06235578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06307095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06035874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06235569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06120613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06274858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06240918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05963735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05963734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05963728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010251 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10561048 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561050 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561051 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561049 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451610_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10561047 | - | - |
| Mouse Genome 430 2.0 Array | 1451610_at | - | - |
| Mouse Genome 430 2.0 Array | 1451610_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 232983_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 232983_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700022766.mm.1 | TC0700002633.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022766.mm.1 | NM_153576.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022766.mm.1 | NM_153576.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022765.mm.1 | TC0700002633.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022765.mm.1 | NM_153576.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022765.mm.1 | NM_153576.2 | Refseq |
| Mouse Expression Set 430 | 1451610_PM_at | - | - |
| Mouse Expression Set 430 | 1451610_at | - | - |
| Murine Genome U74v2 Set | 169733_at | - | - |
| Murine Genome U74v2 Set | 169733_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451610_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451610_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451610_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451610_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6110600 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6110600 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006110600 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226394 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226394 | - | - |
| mouse-6 v1.1 (Illumina) | scl31630.3.1_12-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6110600 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31630.3.1_12-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6110600 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2783414 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1226394 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226394 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2783414 | - | - |

Cxcl17
Genomic Map