Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700004826.mm.1 |
TC0700000559.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700004826.mm.1 |
NM_153577.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700004826.mm.1 |
NM_153577.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700004823.mm.1 |
TC0700000559.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700004823.mm.1 |
NM_153577.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700004823.mm.1 |
NM_153577.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700004817.mm.1 |
TC0700000559.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700004817.mm.1 |
NM_153577.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700004817.mm.1 |
NM_153577.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700004824.mm.1 |
TC0700000559.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700004824.mm.1 |
NM_153577.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700004824.mm.1 |
NM_153577.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700004827.mm.1 |
TC0700000559.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700004827.mm.1 |
NM_153577.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700004827.mm.1 |
NM_153577.2 |
Refseq |