Assay Details
| Gene Symbol: Sv2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109753.1 | NP_001103223.1 | 11 - 12 | 2277 | 79 | |
| NM_153579.4 | NP_705807.2 | 11 - 12 | 2484 | 79 | ||
| GenBank mRNA | AK029736.1 | - | 12 - 13 | 2309 | 79 | |
| AK043830.1 | - | 11 - 12 | 2204 | 79 | ||
| AK122359.1 | - | 10 - 11 | 1429 | 79 | ||
| AK139478.1 | - | 11 - 12 | 2179 | 79 | ||
| AK140104.1 | - | 11 - 12 | 2197 | 79 | ||
| AK147261.1 | - | 11 - 12 | 2271 | 79 | ||
| AK159028.1 | - | 11 - 12 | 2182 | 79 | ||
| BC034651.1 | - | 1 - 2 | 213 | 79 | ||
| BC060224.1 | - | 10 - 11 | 1868 | 30356990 | 79 | |
| BC079568.1 | - | 11 - 12 | 2484 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sv2b
Genomic Map