Assay Details
| Gene Symbol: Atg4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153583.10 | NP_705811.8 | 9 - 10 | 1445 | 77 | |
| GenBank mRNA | AK036282.1 | - | 9 - 10 | 1433 | 77 | |
| AK036571.1 | - | 9 - 10 | 1434 | 77 | ||
| AK081002.1 | - | 9 - 10 | 1432 | 77 | ||
| AK081605.1 | - | 9 - 10 | 1423 | 77 | ||
| AK087258.1 | - | 9 - 10 | 1637 | 77 | ||
| AK154130.1 | - | 9 - 10 | 1438 | 77 | ||
| BC030861.1 | - | 9 - 10 | 1422 | 4208700 | 77 | |
| BC069851.1 | - | 9 - 10 | 1397 | 6490007 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
autophagosome assembly
mitophagy
C-terminal protein lipidation
proteolysis
protein targeting to membrane
transport
autophagy
apoptotic process
cellular response to nitrogen starvation
protein transport
protein processing
nucleophagy
protein delipidation
cysteine-type endopeptidase activity
peptidase activity
cysteine-type peptidase activity
hydrolase activity
autophagosome assembly
mitophagy
C-terminal protein lipidation
proteolysis
protein targeting to membrane
transport
autophagy
apoptotic process
cellular response to nitrogen starvation
protein transport
protein processing
nucleophagy
protein delipidation
cysteine-type endopeptidase activity
peptidase activity
cysteine-type peptidase activity
hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346226 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346230 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346227 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346229 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346228 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346225 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1961135 | BC030861.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1961135 | BC069851.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1961135 | AK154130.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1961135 | AK215761.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1961135 | AK036571.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874278 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 198400 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537503 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510176 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 536475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 51064 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162938 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150466 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1961135 | BC030861.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1961135 | BC069851.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1961135 | AK154130.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1961135 | AK215761.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1961135 | AK036571.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P207106 | AK044798.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P207106 | AK080353.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P418624 | AK036571.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P418624 | NM_153583.9 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P418624 | AK036282.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346226 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01881824 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150466 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06718200 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162938 | - | - |
| Whole Mouse Genome G4122A | A_52_P526750 | BC030861.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P526750 | BC069851.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P526750 | AK081605.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P526750 | AK087258.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P526750 | AK154130.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P207106 | AK044798.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P207106 | AK080353.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P418624 | AK036571.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P418624 | NM_153583.9 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P418624 | AK036282.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121146 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114433 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150466 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162938 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08037584 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08037590 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08037587 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08037582 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346226 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346228 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346230 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346229 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346225 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07459146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07844674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07647106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08011366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08037584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07743424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07939390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07865022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08037582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07550368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07657739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07494687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08107805 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10583575 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583581 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583580 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583577 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583574 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583582 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583583 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583584 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583579 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583576 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583578 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433918_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439130_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10591497 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10583573 | - | - |
| Mouse Genome 430 2.0 Array | 1439130_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433918_at | - | - |
| Mouse Genome 430 2.0 Array | 1439130_at | - | - |
| Mouse Genome 430 2.0 Array | 1433918_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 235040_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 235040_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900001289.mm.1 | NM_153583.10 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001289.mm.1 | TC0900000191.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900001287.mm.1 | NM_153583.10 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001287.mm.1 | TC0900000191.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900001288.mm.1 | NM_153583.10 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001288.mm.1 | TC0900000191.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1433918_PM_at | - | - |
| Mouse Expression Set 430 | 1433918_at | - | - |
| Mouse Expression Set 430 | 1439130_PM_at | - | - |
| Mouse Expression Set 430 | 1439130_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433918_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433918_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433918_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433918_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439130_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439130_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 434918 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106840673 | - | - |
| MouseWG-6 v2 Expression BeadChip | 3290176 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106840673 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106650100 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3290176 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106400086 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101500400 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003290176 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2601710 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2449193 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2601710 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2581779 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237020 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231100 | - | - |
| mouse-6 v1.1 (Illumina) | scl37228.10_481-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00235040.2_157-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3290176 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106400086 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106650100 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106840673 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101500400 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00235040.2_157-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101500400 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3290176 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106650100 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106400086 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106840673 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2601710 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231100 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1235412 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2601710 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237020 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2449193 | - | - |

Atg4d
Genomic Map