Assay Details
| Gene Symbol: Cnot10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153585.5 | NP_705813.2 | 2 - 3 | 381 | 94 | |
| GenBank mRNA | AK029103.1 | - | 2 - 3 | 256 | 94 | |
| AK080503.1 | - | 2 - 3 | 268 | 94 | ||
| AK149944.1 | - | 2 - 3 | 214 | 94 | ||
| AK157824.1 | - | 2 - 3 | 280 | 94 | ||
| AK161842.1 | - | 2 - 3 | 365 | 94 | ||
| AK167288.1 | - | 2 - 3 | 228 | 94 | ||
| AK171701.1 | - | 2 - 3 | 361 | 94 | ||
| BC025554.1 | - | 2 - 3 | 242 | 5321869 | 94 | |
| BC036179.1 | - | 2 - 3 | 353 | 5059723 | 94 | |
| BC039183.1 | - | 2 - 3 | 355 | 2631793 | 94 | |
| CA537067.1 | - | 2 - 3 | 381 | 94 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK167288.1 transcripts.
Set Membership: |
|

Cnot10
Genomic Map