Assay Details
| Gene Symbol: Cnot10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153585.5 | NP_705813.2 | 10 - 11 | 1469 | 101 | |
| GenBank mRNA | AK029103.1 | - | 10 - 11 | 1344 | 101 | |
| AK149944.1 | - | 10 - 11 | 1302 | 101 | ||
| AK157824.1 | - | 10 - 11 | 1368 | 101 | ||
| AK195804.1 | - | 1 - 2 | 89 | 101 | ||
| BC025554.1 | - | 10 - 11 | 1330 | 5321869 | 101 | |
| BC036178.1 | - | 7 - 8 | 925 | 101 | ||
| BC036179.1 | - | 10 - 11 | 1441 | 5059723 | 101 | |
| BC039183.1 | - | 10 - 11 | 1443 | 2631793 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnot10
Genomic Map