Assay Details
| Gene Symbol: Mkl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122667.2 | NP_001116139.1 | 10 - 11 | 1267 | 152 | |
| NM_153588.3 | NP_705816.2 | 8 - 9 | 1281 | 152 | ||
| GenBank mRNA | AF532598.1 | - | 8 - 9 | 1063 | 152 | |
| AK220377.1 | - | 8 - 9 | 1063 | 152 | ||
| BC131644.1 | - | 8 - 9 | 1063 | 40110019 | 152 | |
| BC131645.1 | - | 8 - 9 | 1061 | 152 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mkl2
Genomic Map