Assay Details
| Gene Symbol: Mkl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122667.2 | NP_001116139.1 | 15 - 16 | 2849 | 80 | |
| NM_153588.3 | NP_705816.2 | 13 - 14 | 2863 | 80 | ||
| GenBank mRNA | AF532598.1 | - | 13 - 14 | 2645 | 80 | |
| AK220377.1 | - | 13 - 14 | 2645 | 80 | ||
| BC131644.1 | - | 13 - 14 | 2645 | 40110019 | 80 | |
| BC131645.1 | - | 13 - 14 | 2643 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mkl2
Genomic Map