Assay Details
| Gene Symbol: Klre1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153590.3 | NP_705818.3 | 4 - 5 | 422 | 120 | |
| GenBank mRNA | AF306663.1 | - | 4 - 5 | 422 | 120 | |
| AY100458.1 | - | 4 - 5 | 815 | 120 | ||
| BC145177.1 | - | 4 - 5 | 594 | 120 | ||
More Information
Set Membership: |
|

Klre1
Genomic Map