Assay Details
| Gene Symbol: Tmem17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153596.3 | NP_705824.1 | 1 - 2 | 302 | 55 | |
| GenBank mRNA | BC030386.1 | - | 1 - 2 | 303 | 5400918 | 55 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27641464 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27641466 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27641465 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27641463 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27641461 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27641462 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P152631 | BC030386.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P152631 | NM_153596.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P152631 | BC030386.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P152631 | NM_153596.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80992 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 28288 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 366436 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580110 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 146426 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164855 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P152631 | BC030386.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P152631 | NM_153596.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P152631 | BC030386.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P152631 | NM_153596.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641472 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27641467 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164855 | - | - |
| Whole Mouse Genome G4122A | A_52_P152631 | BC030386.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P152631 | NM_153596.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P152631 | BC030386.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P152631 | NM_153596.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P488014 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128198 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164855 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09029866 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09029856 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27641461 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27641463 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27641462 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27641466 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27641465 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27641464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27641453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09651331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09010052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09443120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09305848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09440793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27641457 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10374555 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10374557 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10374554 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10374556 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459997_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456332_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10374553 | - | - |
| Mouse Genome 430 2.0 Array | 1456332_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456332_at | - | - |
| Mouse Genome 430 2.0 Array | 1459997_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1459997_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 103765_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 103765_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100001825.mm.1 | NM_153596.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100001825.mm.1 | NM_153596.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100001825.mm.1 | TC1100000223.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100001826.mm.1 | NM_153596.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100001826.mm.1 | NM_153596.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100001826.mm.1 | TC1100000223.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456332_at | - | - |
| Mouse Expression Set 430 | 1459997_PM_s_at | - | - |
| Mouse Expression Set 430 | 1456332_PM_at | - | - |
| Mouse Expression Set 430 | 1459997_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456332_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459997_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459997_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456332_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 769956 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5900079 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5900079 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005900079 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231660 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231660 | - | - |
| mouse-6 v1.1 (Illumina) | scl41791.4.1_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5900079 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41791.4.1_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5900079 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2907721 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1231660 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231660 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2907721 | - | - |

Tmem17
Genomic Map