Assay Details
| Gene Symbol: Tmem17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153596.3 | NP_705824.1 | 2 - 3 | 404 | 84 | |
| GenBank mRNA | BC030386.1 | - | 2 - 3 | 405 | 5400918 | 84 |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem17
Genomic Map