Assay Details
| Gene Symbol: Trpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153597.2 | NP_705825.2 | 5 - 6 | 882 | 66 | |
| NR_027351.1 | 4 - 5 | 712 | 66 | |||
| GenBank mRNA | AK038830.1 | - | 4 - 5 | 714 | 66 | |
| AK048558.1 | - | 1031 | 66 | |||
| AK090183.1 | - | 4 - 5 | 2013 | 66 | ||
| AK141582.1 | - | 669 | 66 | |||
| BC027575.1 | - | 5 - 6 | 680 | 1364225 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trpt1
Genomic Map