Assay Details
| Gene Symbol: Lrrc56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172064.1 | NP_001165535.1 | 3 - 4 | 745 | 68 | |
| NM_001172065.1 | NP_001165536.1 | 3 - 4 | 745 | 68 | ||
| NM_153777.2 | NP_722472.1 | 3 - 4 | 331 | 68 | ||
| GenBank mRNA | AK017603.1 | - | 3 - 4 | 719 | 68 | |
| AK220237.1 | - | 3 - 4 | 699 | 68 | ||
| BC027807.1 | - | 3 - 4 | 341 | 5324022 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc56
Genomic Map