Assay Details
| Gene Symbol: Acap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153788.3 | NP_722483.2 | 9 - 10 | 945 | 114 | |
| GenBank mRNA | AK138736.1 | - | 9 - 10 | 938 | 114 | |
| AK154210.1 | - | 9 - 10 | 910 | 115 | ||
| AK154449.1 | - | 9 - 10 | 948 | 114 | ||
| AK155001.1 | - | 9 - 10 | 966 | 114 | ||
| BC037481.1 | - | 4 - 5 | 572 | 4460639 | 114 | |
More Information
Set Membership: |
|

Acap1
Genomic Map