Assay Details
| Gene Symbol: Tecrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153801.3 | NP_722496.2 | 7 - 8 | 923 | 114 | |
| GenBank mRNA | AF548365.1 | - | 7 - 8 | 924 | 114 | |
| AK052276.1 | - | 7 - 8 | 762 | 114 | ||
| AK052284.1 | - | 7 - 8 | 926 | 114 | ||
| AK052302.1 | - | 7 - 8 | 628 | 114 | ||
| AK052378.1 | - | 7 - 8 | 686 | 114 | ||
| BC107366.1 | - | 7 - 8 | 836 | 40056219 | 114 | |
| BC107367.1 | - | 7 - 8 | 836 | 40056223 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tecrl
Genomic Map