| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200004051.mm.1 |
NM_153805.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004051.mm.1 |
NM_153805.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004051.mm.1 |
TC0200000523.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200004052.mm.1 |
NM_153805.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004052.mm.1 |
NM_153805.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004052.mm.1 |
TC0200000523.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200004053.mm.1 |
NM_153805.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004053.mm.1 |
NM_153805.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004053.mm.1 |
TC0200000523.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200004054.mm.1 |
NM_153805.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004054.mm.1 |
NM_153805.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200004054.mm.1 |
TC0200000523.mm.1 |
MTA TC |