Assay Details
| Gene Symbol: Pkn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153805.1 | NP_722500.1 | 6 - 7 | 825 | 78 | |
| GenBank mRNA | AK142182.1 | - | 5 - 6 | 1341 | 78 | |
| AK142988.1 | - | 5 - 6 | 1397 | 78 | ||
| AK154801.1 | - | 5 - 6 | 1363 | 78 | ||
| BC034126.1 | - | 6 - 7 | 825 | 4912536 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pkn3
Genomic Map