Assay Details
| Gene Symbol: Pkn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153805.1 | NP_722500.1 | 9 - 10 | 1227 | 88 | |
| GenBank mRNA | AK142182.1 | - | 8 - 9 | 1743 | 88 | |
| AK142988.1 | - | 8 - 9 | 1799 | 88 | ||
| AK154801.1 | - | 8 - 9 | 1765 | 88 | ||
| BC034126.1 | - | 9 - 10 | 1227 | 4912536 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pkn3
Genomic Map