Assay Details
| Gene Symbol: Igsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170599.2 | NP_733548.2 | 2 - 3 | 381 | 64 | |
| GenBank mRNA | AB079880.1 | - | 2 - 3 | 218 | 64 | |
| AK160146.1 | - | 2 - 3 | 344 | 64 | ||
| BC057555.1 | - | 2 - 3 | 381 | 6416190 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igsf11
Genomic Map