Assay Details
| Gene Symbol: Kcnn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163510.1 | NP_001156982.1 | 2 - 3 | 426 | 75 | |
| NM_008433.4 | NP_032459.3 | 2 - 3 | 426 | 75 | ||
| GenBank mRNA | AF042487.1 | - | 2 - 3 | 292 | 75 | |
| AF072884.1 | - | 2 - 3 | 252 | 75 | ||
| AK010943.1 | - | 2 - 3 | 427 | 75 | ||
| AK085752.1 | - | 2 - 3 | 236 | 75 | ||
| AK155910.1 | - | 2 - 3 | 391 | 75 | ||
| BC010274.1 | - | 2 - 3 | 375 | 3589411 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnn4
Genomic Map