Assay Details
| Gene Symbol: Kcnn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163510.1 | NP_001156982.1 | 6 - 7 | 1218 | 71 | |
| NM_008433.4 | NP_032459.3 | 6 - 7 | 1218 | 71 | ||
| GenBank mRNA | AB128930.1 | - | 4 - 5 | 981 | 71 | |
| AF042487.1 | - | 6 - 7 | 1084 | 71 | ||
| AF072884.1 | - | 6 - 7 | 1044 | 71 | ||
| AK010943.1 | - | 6 - 7 | 1219 | 71 | ||
| AK085752.1 | - | 6 - 7 | 1035 | 71 | ||
| AK088778.1 | - | 1 - 2 | 142 | 71 | ||
| AK155910.1 | - | 6 - 7 | 1183 | 71 | ||
| BC010274.1 | - | 6 - 7 | 1167 | 3589411 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnn4
Genomic Map