Assay Details
| Gene Symbol: Nsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008739.3 | NP_032765.3 | 12 - 13 | 4883 | 76 | |
| GenBank mRNA | BQ442794.1 | - | 5 - 6 | 554 | 76 | |
| CB247038.1 | - | 4 - 5 | 434 | 76 | ||
More Information
Set Membership: |
|

Nsd1
Genomic Map