Assay Details
| Gene Symbol: Psmc3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008949.3 | NP_032975.1 | 2 - 3 | 198 | 69 | |
| GenBank mRNA | AB000121.1 | - | 2 - 3 | 163 | 69 | |
| AK133462.1 | - | 2 - 3 | 166 | 69 | ||
| BC030169.1 | - | 2 - 3 | 175 | 5027493 | 69 | |
| FJ937966.1 | - | 2 - 3 | 169 | 69 | ||
| FJ937967.1 | - | 1 - 2 | 233 | 69 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the FJ937967.1 transcripts.
Set Membership: |
|

Psmc3ip
Genomic Map