Assay Details
| Gene Symbol: Psmc3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008949.3 | NP_032975.1 | 3 - 4 | 284 | 66 | |
| GenBank mRNA | AB000121.1 | - | 3 - 4 | 249 | 66 | |
| BC030169.1 | - | 3 - 4 | 261 | 5027493 | 66 | |
| FJ937966.1 | - | 3 - 4 | 255 | 66 | ||
| FJ937967.1 | - | 2 - 3 | 319 | 66 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the FJ937967.1 transcripts.
Set Membership: |
|

Psmc3ip
Genomic Map