Assay Details
| Gene Symbol: Gnpat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010322.3 | NP_034452.3 | 4 - 5 | 770 | 62 | |
| GenBank mRNA | AF110769.1 | - | 4 - 5 | 734 | 62 | |
| AJ132012.1 | - | 4 - 5 | 565 | 62 | ||
| AK004630.1 | - | 4 - 5 | 744 | 62 | ||
| AK004771.1 | - | 4 - 5 | 750 | 62 | ||
| AK010896.1 | - | 4 - 5 | 744 | 62 | ||
| AK166997.1 | - | 4 - 5 | 742 | 62 | ||
| AK215406.1 | - | 1 - 2 | 82 | 62 | ||
| BC003225.1 | - | 4 - 5 | 712 | 62 | ||
| BC025972.1 | - | 4 - 5 | 727 | 4020505 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnpat
Genomic Map