Assay Details
| Gene Symbol: Mecp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010788.4 | NP_034918.1 | 2 - 3 | 309 | 64 | |
| GenBank mRNA | AF158181.1 | - | 2 - 3 | 212 | 64 | |
| AJ132922.1 | - | 1 - 2 | 65 | 64 | ||
| BC027153.1 | - | 2 - 3 | 231 | 4948925 | 64 | |
| BY107013.1 | - | 2 - 3 | 257 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF158181.1, AJ132922.1, BC027153.1, NM_010788.4 transcripts.
Set Membership: |
|

Mecp2
Genomic Map