Assay Details
| Gene Symbol: Prkdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011159.2 | NP_035289.2 | 55 - 56 | 7463 | 149 | |
| GenBank mRNA | AB007544.1 | - | 55 - 56 | 7440 | 149 | |
| D87521.1 | - | 55 - 56 | 7440 | 149 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkdc
Genomic Map