Assay Details
| Gene Symbol: Prkdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011159.2 | NP_035289.2 | 63 - 64 | 8806 | 69 | |
| GenBank mRNA | AB007544.1 | - | 63 - 64 | 8783 | 69 | |
| AB011543.1 | - | 63 - 64 | 8795 | 69 | ||
| D87521.1 | - | 63 - 64 | 8783 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkdc
Genomic Map