Assay Details
| Gene Symbol: Kcnh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013569.2 | NP_038597.2 | 4 - 5 | 1242 | 130 | |
| GenBank mRNA | AF012868.1 | - | 4 - 5 | 1248 | 130 | |
| AY374424.1 | - | 4 - 5 | 927 | 130 | ||
| BC051016.1 | - | 4 - 5 | 1073 | 130 | ||
| BQ745654.1 | - | 1 - 2 | 136 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnh2
Genomic Map