Assay Details
| Gene Symbol: Kcnh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294162.1 | NP_001281091.1 | 8 - 9 | 2270 | 130 | |
| NM_013569.2 | NP_038597.2 | 12 - 13 | 3298 | 130 | ||
| GenBank mRNA | AF012868.1 | - | 12 - 13 | 3304 | 130 | |
| AF012869.1 | - | 8 - 9 | 2218 | 130 | ||
| AF034762.1 | - | 8 - 9 | 2262 | 130 | ||
| AY374424.1 | - | 12 - 13 | 2983 | 130 | ||
| BC051016.1 | - | 12 - 13 | 3129 | 130 | ||
| BC051233.1 | - | 2 - 3 | 336 | 130 | ||
| CK619061.1 | - | 3 - 4 | 525 | 130 | ||
| CX565252.1 | - | 1 - 2 | 313 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnh2
Genomic Map